Learn More
BioVendor, LLC Polyclonal AntibodyWestern blotting, ELISA,
SDP
Supplier:  BioVendor, LLC RD181060100
Encompass
Polyclonal AntibodyWestern blotting, ELISA, Immunohistochemistry Source of AntigenE. coliHostsRabbitPreparationThe antibody was raised in rabbits by immunization with the recombinant Human FABP5.Amino Acid SequenceThe immunization antigen (15.2 kDa) is a protein containing 135 AA of recombinant Human FABP5 and one extra AA, N-terminal methionin (highlighted).MATVQQLEGRWRLVDSKGFDEYMKELGVGIALRKMGAMAKPDCIITCDGKNLTIKTESTLKTTQFSCTLGEKFEETTADGRKTQTVCNFTDGALVQHQEWDGKESTITRKLKDGKLVVECVMNNVTCTRIYEKVESpecies ReactivityHuman. Not yet tested in other species.Purification MethodImmunoaffinity chromatography on a column with immobilized recombinant Human FABP5.Antibody Content0.1 mg (determined by BCA method, BSA was used as a standard
Catalog No. 50-894-078