Learn More
Cayman Chemical Falcarinol, Purity: ≥60%, CAS: 21852-80-2, 5 mg
SDP
Catalog No. 502986025
Encompass_Preferred
A plant defense polyacetylene; inhibits fatty acid biosynthesis in pathogens; binds to hCB1 and hCB2 receptors (Kis = 594 and 2,100 nM, respectively); modulates synaptic and extrasynaptic GABAA receptors; inhibits ATPase activity in ABCG2 (IC50 = 79.3 μM); inhibits aberrant crypt foci (26.6%) in azoxymethane-induced rats
Catalog No. 50-298-6025 Supplier Cayman Chemical Supplier No. 224075 MG
A plant defense polyacetylene; inhibits fatty acid biosynthesis in pathogens; binds to hCB1 and hCB2 receptors (Kis = 594 and 2,100 nM, respectively); modulates synaptic and extrasynaptic GABAA receptors; inhibits ATPase activity in ABCG2 (IC50 = 79.3 μM); inhibits aberrant crypt foci (26.6%) in azoxymethane-induced rats