Learn More
Cayman Chemical ANTIBODY Famoxadon 50mg
SDP
Supplier:  Cayman Chemical 3447050 MG
Encompass_Preferred
A fungicide; inhibits mitochondrial complex III and electron transport in rat heart submitochondria (IC50s = 28 and 4.5 ng/ml, respectively); reduces oxygen consumption in, and induces disintegration of, P. infestans zoospores at 2 UG/ml; inhibits the growth of P. viticola on grape seedlings (EC50 = 3 UG/ml)
Catalog No. 50-294-8522