Learn More
Cayman Chemical FDU-PB-22, Purity: ≥98%, CAS: 1883284-94-3, 10 mg
SDP
Catalog No. 502973299
Encompass_Preferred
An analytical reference standard categorized as a synthetic cannabinoid; has been found in samples seized by law enforcement; intended for research and forensic applications
Catalog No. 50-297-3299 Supplier Cayman Chemical Supplier No. 1496810 MG
An analytical reference standard categorized as a synthetic cannabinoid; has been found in samples seized by law enforcement; intended for research and forensic applications