Learn More
Cayman Chemical FF-MAS, Purity: ≥90%, CAS: 64284-64-6, 1 mg
SDP
Catalog No. 502932724
Encompass_Preferred
A sterol; an intermediate in cholesterol biosynthesis from lanosterol; found at high concentrations in mammalian ovarian follicular fluid; increases survival of human oocytes isolated from patients with PCOS; improves cytoplasmic maturation of immature rodent and porcine oocytes in vitro; promotes completion of meiotic maturation to metaphase II and increases the rate of successful IVF in isolated mouse oocytes
Catalog No. 50-293-2724 Supplier Cayman Chemical Supplier No. 260931 MG
A sterol; an intermediate in cholesterol biosynthesis from lanosterol; found at high concentrations in mammalian ovarian follicular fluid; increases survival of human oocytes isolated from patients with PCOS; improves cytoplasmic maturation of immature rodent and porcine oocytes in vitro; promotes completion of meiotic maturation to metaphase II and increases the rate of successful IVF in isolated mouse oocytes