Learn More
Cayman Chemical FIN56, Purity: ≥98%, CAS: 1083162-61-1, 5 mg
SDP
Catalog No. 502933864
Encompass_Preferred
An inducer of ferroptosis; induces ferroptosis in BJ cells overexpressing oncogenic HRASG12V via acetyl-CoA carboxylase-dependent reduction in expression of GPX4 protein; binds to and activates squalene synthase in a GPX4 degradation-independent manner
Catalog No. 50-293-3864 Supplier Cayman Chemical Supplier No. 251805 MG
An inducer of ferroptosis; induces ferroptosis in BJ cells overexpressing oncogenic HRASG12V via acetyl-CoA carboxylase-dependent reduction in expression of GPX4 protein; binds to and activates squalene synthase in a GPX4 degradation-independent manner