Learn More
Cayman Chemical FK-3311, Purity: ≥98%, CAS: 116686-15-8, 1 mg
SDP
Catalog No. 502980040
Encompass_Preferred
A COX-2 inhibitor; inhibits LPS-induced TxB2 production (IC50 = 316 nM) in vitro; inhibits adjuvant-induced arthritis (ED50 = 0.29 mg/kg) in rats without causing gastrointestinal irritation; alleviates ischemia-repurfusion injury in canine liver and rat lung in vivo
Catalog No. 50-298-0040 Supplier Cayman Chemical Supplier No. 176971 MG
A COX-2 inhibitor; inhibits LPS-induced TxB2 production (IC50 = 316 nM) in vitro; inhibits adjuvant-induced arthritis (ED50 = 0.29 mg/kg) in rats without causing gastrointestinal irritation; alleviates ischemia-repurfusion injury in canine liver and rat lung in vivo