Learn More
Cayman Chemical Fluindione, Purity: ≥98%, CAS: 957-56-2, 10 mg
SDP
Catalog No. 502910423
Encompass_Preferred
A vitamin K antagonist; inhibits VKOR (IC50 = 6.6 nM), including VKOR containing certain substitution mutations; inhibits platelet and fibrin deposition, as well as prevents β-thromboglobulin release and thrombin-antithrombin complex formation, in an ex vivo model of tissue factor-induced thrombus formation using isolated human whole blood
Catalog No. 50-291-0423 Supplier Cayman Chemical Supplier No. 3496610 MG
A vitamin K antagonist; inhibits VKOR (IC50 = 6.6 nM), including VKOR containing certain substitution mutations; inhibits platelet and fibrin deposition, as well as prevents β-thromboglobulin release and thrombin-antithrombin complex formation, in an ex vivo model of tissue factor-induced thrombus formation using isolated human whole blood