Learn More
Cayman Chemical Flumequine, Purity: ≥98%, CAS: 42835-25-6, 10 g
SDP
Catalog No. 502986065
Encompass_Preferred
A fluoroquinolone antibiotic; active against S. aureus, S. pyogenes, B. subtilis, E. coli, P. aeruginosa, S. faecalis, and K. pneumoniae (MICs = 1-100 µg/ml); active against field isolates of B. hyodysenteriae (MICs = 6.25-200 µg/ml); increases survival in rat models of P. vulgaris-induced urinary tract infection and P. mirabilis-induced prostatitis at 50 mg/kg
Catalog No. 50-298-6065 Supplier Cayman Chemical Supplier No. 2164510 G
A fluoroquinolone antibiotic; active against S. aureus, S. pyogenes, B. subtilis, E. coli, P. aeruginosa, S. faecalis, and K. pneumoniae (MICs = 1-100 µg/ml); active against field isolates of B. hyodysenteriae (MICs = 6.25-200 µg/ml); increases survival in rat models of P. vulgaris-induced urinary tract infection and P. mirabilis-induced prostatitis at 50 mg/kg