Learn More
Cayman Chemical Flutamide, Purity: ≥98%, CAS: 13311-84-7, 25 g
SDP
Catalog No. 502982968
Encompass_Preferred
An androgen receptor antagonist (Kis = 1.3 and 7.5 µM for the cytosolic rat ventral prostate and anterior pituitary receptors, respectively) and a prodrug form of 2-hydroxy flutamide; cytotoxic to PC3 and LNCaP prostate cancer cells (IC50s = 98.8 and 81.8 µM, respectively); reduces tumor growth in a PC-82 mouse xenograft model at 50 mg/kg per day
Catalog No. 50-298-2968 Supplier Cayman Chemical Supplier No. 2035925 G
An androgen receptor antagonist (Kis = 1.3 and 7.5 µM for the cytosolic rat ventral prostate and anterior pituitary receptors, respectively) and a prodrug form of 2-hydroxy flutamide; cytotoxic to PC3 and LNCaP prostate cancer cells (IC50s = 98.8 and 81.8 µM, respectively); reduces tumor growth in a PC-82 mouse xenograft model at 50 mg/kg per day