Learn More
Cayman Chemical Forskolin, Purity: ≥98%, CAS: 66575-29-9, 10 mg
SDP
Catalog No. 502952133
Encompass_Preferred
A naturally occurring diterpene that directly activates adenylyl cyclase to raise levels of cAMP in a wide variety of cell types; binds to type 1 adenylyl cyclase membranes with an IC50 value of 41 nM and demonstrates an EC50 value of 0.5 μM in an activation assay
Catalog No. 50-295-2133 Supplier Cayman Chemical Supplier No. 1101810 MG
A naturally occurring diterpene that directly activates adenylyl cyclase to raise levels of cAMP in a wide variety of cell types; binds to type 1 adenylyl cyclase membranes with an IC50 value of 41 nM and demonstrates an EC50 value of 0.5 μM in an activation assay