Learn More
Cayman Chemical FRAX597, Purity: ≥98%, CAS: 1286739-19-2, 25 mg
SDP
Catalog No. 502986038
Encompass_Preferred
A PAK inhibitor selective for group I PAKs (IC50s = 7.7, 12.8, and 19.3 nM for PAK 1, 2, and 3, respectively) over group II PAKs (IC50 > 10 µM for PAK4 and 50 = 70 nM); reduces proliferation and survival of transformed Schwann cells, ovarian, and pancreatic cells; works synergistically with gemcitabine in pancreatic cancer models in vitro and in vivo
Catalog No. 50-298-6038 Supplier Cayman Chemical Supplier No. 2220525 MG
A PAK inhibitor selective for group I PAKs (IC50s = 7.7, 12.8, and 19.3 nM for PAK 1, 2, and 3, respectively) over group II PAKs (IC50 > 10 µM for PAK4 and 50 = 70 nM); reduces proliferation and survival of transformed Schwann cells, ovarian, and pancreatic cells; works synergistically with gemcitabine in pancreatic cancer models in vitro and in vivo