Learn More
Cayman Chemical Fz25, Purity: ≥98%, CAS: 2762498-70-2, 50 mg
SDP
Catalog No. 502917705
Encompass_Preferred
A triazole-modified sterol and an anticancer agent; inhibits the proliferation of A2780 and HT-29 cells (IC50s = 15.29 and 15.98 µM, respectively); decreases the phosphorylation of EGFR and STAT3 in A2780 cells at 30.58 µM and phosphorylation of ERK, STAT3, and mTOR in HT-29 cells at 31.96 µM; reduces P-gp efflux activity in a reporter assay using HEK293 cells at 10 µM
Catalog No. 50-291-7705 Supplier Cayman Chemical Supplier No. 3775250 MG
A triazole-modified sterol and an anticancer agent; inhibits the proliferation of A2780 and HT-29 cells (IC50s = 15.29 and 15.98 µM, respectively); decreases the phosphorylation of EGFR and STAT3 in A2780 cells at 30.58 µM and phosphorylation of ERK, STAT3, and mTOR in HT-29 cells at 31.96 µM; reduces P-gp efflux activity in a reporter assay using HEK293 cells at 10 µM