Learn More
Cayman Chemical GBT-440, Purity: ≥98%, CAS: 1446321-46-5, 5 mg
SDP
Catalog No. 502994349
Encompass_Preferred
A modulator of sickle hemoglobin; increases HbS affinity for oxygen (EC50s = 8.2 and 415 μM for purified HbS and whole blood from sickle cell anemia patients, respectively); delays oxygen release from and polymerization of HbS in a concentration-dependent manner; increases red blood cell half-life and reduces sickling ex vivo in whole blood isolated from a murine model of sickle cell anemia
Catalog No. 50-299-4349 Supplier Cayman Chemical Supplier No. 239335 MG
A modulator of sickle hemoglobin; increases HbS affinity for oxygen (EC50s = 8.2 and 415 μM for purified HbS and whole blood from sickle cell anemia patients, respectively); delays oxygen release from and polymerization of HbS in a concentration-dependent manner; increases red blood cell half-life and reduces sickling ex vivo in whole blood isolated from a murine model of sickle cell anemia