Learn More
Cayman Chemical GCN2-IN-1, Purity: ≥90%, CAS: 1448693-69-3, 25 mg
SDP
Catalog No. 502911919
Encompass_Preferred
An inhibitor of GCN2 (IC50 = ~0.2 µM for autophosphorylation); inhibits integrase phosphorylation (IC50 = ~0.2 µM for the HIV-1 enzyme); inhibits ELAV-like protein 1 protein dimerization (IC50 = 4.5 µM); decreases the viability in U251, XD456, U87, and LN-229 glioma cells (IC50s = 4.7, 2.8, 3.2, and 2.7 µM, respectively)
Catalog No. 50-291-1919 Supplier Cayman Chemical Supplier No. 3602725 MG
An inhibitor of GCN2 (IC50 = ~0.2 µM for autophosphorylation); inhibits integrase phosphorylation (IC50 = ~0.2 µM for the HIV-1 enzyme); inhibits ELAV-like protein 1 protein dimerization (IC50 = 4.5 µM); decreases the viability in U251, XD456, U87, and LN-229 glioma cells (IC50s = 4.7, 2.8, 3.2, and 2.7 µM, respectively)