Learn More
Cayman Chemical GIT 27, Purity: ≥98%, CAS: 6501-72-0, 50 mg
SDP
Catalog No. 502970497
Encompass_Preferred
An immunomodulatory compound that inhibits inflammatory antigen presentation both in vitro and in preclinical in vivo models of autoimmune diseases; targets macrophages, reducing the production of the proinflammatory mediators TNF-α, IL-1β, macrophage migration inhibitory factor, and inducible nitric oxide synthase-mediated nitric oxide generation
Catalog No. 50-297-0497 Supplier Cayman Chemical Supplier No. 1629350 MG
An immunomodulatory compound that inhibits inflammatory antigen presentation both in vitro and in preclinical in vivo models of autoimmune diseases; targets macrophages, reducing the production of the proinflammatory mediators TNF-α, IL-1β, macrophage migration inhibitory factor, and inducible nitric oxide synthase-mediated nitric oxide generation