Learn More
Cayman Chemical GK187, Purity: ≥95%, CAS: 1071001-50-7, 5 mg
SDP
Catalog No. 502935212
Encompass_Preferred
A GVIA iPLA2 inhibitor; inhibits GVIA iPLA2 by 99.8% at 0.091 mole fraction; selective for GVIA iPLA2 over GIVA cPLA2 and GV sPLA2 where it shows less than 25% and 32.8% inhibition, respectively
Catalog No. 50-293-5212 Supplier Cayman Chemical Supplier No. 251305 MG
A GVIA iPLA2 inhibitor; inhibits GVIA iPLA2 by 99.8% at 0.091 mole fraction; selective for GVIA iPLA2 over GIVA cPLA2 and GV sPLA2 where it shows less than 25% and 32.8% inhibition, respectively