Learn More
Cayman Chemical GK241, Purity: ≥98%, CAS: 1899930-93-8, 10 mg
SDP
Catalog No. 502916281
Encompass_Preferred
An sPLA2 (Type IIA) inhibitor (IC50s = 0.143 and 0.068 µM for the human and mouse enzymes, respectively); inhibits IL-1β-induced production of PGE2 in isolated rat renal mesangial cells at 3 and 10 µM; inhibits SARS-CoV-2 Mpro (IC50 = 24 µM)
Catalog No. 50-291-6281 Supplier Cayman Chemical Supplier No. 3768910 MG
An sPLA2 (Type IIA) inhibitor (IC50s = 0.143 and 0.068 µM for the human and mouse enzymes, respectively); inhibits IL-1β-induced production of PGE2 in isolated rat renal mesangial cells at 3 and 10 µM; inhibits SARS-CoV-2 Mpro (IC50 = 24 µM)