Learn More
Cayman Chemical GKT136901, Purity: ≥95%, CAS: 955272-06-7, 50 mg
SDP
Catalog No. 502946139
Encompass_Preferred
A dual inhibitor of NOX1 and NOX4 (Kis = 160 and 165 nM, respectively); selective for NOX1 and NOX4 over NOX2 and xanthine oxidase (Kis = 1,530 and >30,000 nM, respectively), as well as a panel of 18 enzymes and ion channels at 10 µM; scavenges peroxynitrite radicals, as well as prevents the nitration and dimer formation of α-synuclein, in cell-free assays at 0.1, 1, and 10 µM; prevents peroxynitrite-induced cell death of differentiated LUHMES cells at 10 µM
Catalog No. 50-294-6139 Supplier Cayman Chemical Supplier No. 3420550 MG
A dual inhibitor of NOX1 and NOX4 (Kis = 160 and 165 nM, respectively); selective for NOX1 and NOX4 over NOX2 and xanthine oxidase (Kis = 1,530 and >30,000 nM, respectively), as well as a panel of 18 enzymes and ion channels at 10 µM; scavenges peroxynitrite radicals, as well as prevents the nitration and dimer formation of α-synuclein, in cell-free assays at 0.1, 1, and 10 µM; prevents peroxynitrite-induced cell death of differentiated LUHMES cells at 10 µM