Learn More
Cayman Chemical Glaucine, Purity: ≥95%, CAS: 475-81-0, 10 mg
SDP
Catalog No. 502964920
Encompass_Preferred
An analytical reference standard categorized as an antitussive; has hallucinogenic properties; intended for research and forensic applications
Catalog No. 50-296-4920 Supplier Cayman Chemical Supplier No. 1733810 MG
An analytical reference standard categorized as an antitussive; has hallucinogenic properties; intended for research and forensic applications