Learn More
Cayman Chemical GLPG2938, Purity: ≥98%, CAS: 2130996-00-6, 25 mg
SDP
Catalog No. 502947595
Encompass_Preferred
An S1P2 antagonist; inhibits S1P-induced calcium flux in CHO cells overexpressing human S1P2 (IC50 = 8.8 nM); inhibits S1P-induced IL-8 release in HFL1 cells (IC50 = 0.6 nM) and S1P-induced contraction of human lung fibroblasts; prevents epithelial damage and pulmonary fibrosis, structural distortion, and inflammation in a mouse model of bleomycin-induced pulmonary fibrosis at 3, 10, and 30 mg/kg
Catalog No. 50-294-7595 Supplier Cayman Chemical Supplier No. 3431825 MG
An S1P2 antagonist; inhibits S1P-induced calcium flux in CHO cells overexpressing human S1P2 (IC50 = 8.8 nM); inhibits S1P-induced IL-8 release in HFL1 cells (IC50 = 0.6 nM) and S1P-induced contraction of human lung fibroblasts; prevents epithelial damage and pulmonary fibrosis, structural distortion, and inflammation in a mouse model of bleomycin-induced pulmonary fibrosis at 3, 10, and 30 mg/kg