Learn More
Cayman Chemical Glutaurine, Purity: ≥90%, CAS: 56488-60-9, 50 mg
SDP
Catalog No. 502918310
Encompass_Preferred
A dipeptide; enhances potassium-induced release of L-glutamate and inhibits kainate-induced formation of cGMP in isolated mouse cerebral cortical slices at 0.1 and 1 mM, respectively; decreases plasma levels of T3 and increases plasma TSH levels in rats at 600 µg/kg; prevents electroconvulsive shock-induced amnesia in rats at 50 µg/animal, p.o.
Catalog No. 50-291-8310 Supplier Cayman Chemical Supplier No. 3958650 MG
A dipeptide; enhances potassium-induced release of L-glutamate and inhibits kainate-induced formation of cGMP in isolated mouse cerebral cortical slices at 0.1 and 1 mM, respectively; decreases plasma levels of T3 and increases plasma TSH levels in rats at 600 µg/kg; prevents electroconvulsive shock-induced amnesia in rats at 50 µg/animal, p.o.