Learn More
Cayman Chemical GLX-351322, Purity: ≥98%, CAS: 835598-94-2, 10 mg
SDP
Catalog No. 502990961
Encompass_Preferred
A NOX4 inhibitor (IC50 = 5 µM); selective for NOX4 over NOX2 (IC50 = 40 µM); reduces glucose-induced production of ROS and cytotoxicity in isolated human pancreatic islets at 10 µM; decreases blood glucose levels in an intravenous glucose tolerance test in mice fed a high-fat diet at an estimated 3.8 mg/kg per day
Catalog No. 50-299-0961 Supplier Cayman Chemical Supplier No. 3152310 MG
A NOX4 inhibitor (IC50 = 5 µM); selective for NOX4 over NOX2 (IC50 = 40 µM); reduces glucose-induced production of ROS and cytotoxicity in isolated human pancreatic islets at 10 µM; decreases blood glucose levels in an intravenous glucose tolerance test in mice fed a high-fat diet at an estimated 3.8 mg/kg per day