Learn More
Cayman Chemical GNE-3511, Purity: ≥98%, CAS: 1496581-76-0, 25 mg
SDP
Catalog No. 502982138
Encompass_Preferred
A selective and highly potent inhibitor of DLK; also inhibits phosphorylated JNK (IC50 = 30 nM); selective over other MAP kinases, JNKs, and MLKs; protects primary neurons in an in vitro axon degeneration assay (IC50 = 107 nM); decreases p-c-Jun expression in a mouse MPTP model of Parkinson’s disease
Catalog No. 50-298-2138 Supplier Cayman Chemical Supplier No. 1917425 MG
A selective and highly potent inhibitor of DLK; also inhibits phosphorylated JNK (IC50 = 30 nM); selective over other MAP kinases, JNKs, and MLKs; protects primary neurons in an in vitro axon degeneration assay (IC50 = 107 nM); decreases p-c-Jun expression in a mouse MPTP model of Parkinson’s disease