Learn More
Cayman Chemical GNE-477, Purity: ≥98%, CAS: 1032754-81-6, 50 mg
SDP
Catalog No. 502940774
Encompass_Preferred
A dual PI3K (IC50 = 4 nM for PI3Kα) and mTOR inhibitor (apparent Ki = 21 nM); inhibits proliferation of MCF-7.1 cells (EC50 = 143 nM); reduces tumor growth in a PC3-NCI tumor xenograft model at 1-20 mg/kg per day
Catalog No. 50-294-0774 Supplier Cayman Chemical Supplier No. 2797750 MG
A dual PI3K (IC50 = 4 nM for PI3Kα) and mTOR inhibitor (apparent Ki = 21 nM); inhibits proliferation of MCF-7.1 cells (EC50 = 143 nM); reduces tumor growth in a PC3-NCI tumor xenograft model at 1-20 mg/kg per day