Learn More
Cayman Chemical GRL-0496, Purity: ≥98%, CAS: 1087243-14-8, 1 mg
SDP
Catalog No. 502909190
Encompass_Preferred
An inhibitor of SARS-CoV and SARS-CoV-2 Mpro (IC50s = 0.03 and 0.038 µM, respectively); reduces viral infectivity in SARS-CoV-infected Vero cells (EC50 = 6.9 µM)
Catalog No. 50-290-9190 Supplier Cayman Chemical Supplier No. 361141 MG
An inhibitor of SARS-CoV and SARS-CoV-2 Mpro (IC50s = 0.03 and 0.038 µM, respectively); reduces viral infectivity in SARS-CoV-infected Vero cells (EC50 = 6.9 µM)