Learn More
Cayman Chemical GS-7340, Purity: ≥98%, CAS: 379270-37-8, 100 mg
SDP
Catalog No. 502935156
Encompass_Preferred
A prodrug from of tenofovir; reduces HIV-1 viral replication in MT-2 cells (EC50 = 5 nM)
Catalog No. 50-293-5156 Supplier Cayman Chemical Supplier No. 26214100 MG
A prodrug from of tenofovir; reduces HIV-1 viral replication in MT-2 cells (EC50 = 5 nM)