Learn More
Cayman Chemical GSK2018682, Purity: ≥95%, CAS: 1034688-30-6, 10 mg
SDP
Catalog No. 502991123
Encompass_Preferred
An S1P1 and S1P5 receptor agonist; selective for S1P1 and S1P5 over S1P2, S1P3, and S1P4 receptors; efficacious in a mouse model of EAE
Catalog No. 50-299-1123 Supplier Cayman Chemical Supplier No. 3143710 MG
An S1P1 and S1P5 receptor agonist; selective for S1P1 and S1P5 over S1P2, S1P3, and S1P4 receptors; efficacious in a mouse model of EAE