Learn More
Cayman Chemical GSK2193874, Purity: ≥95%, CAS: 1336960-13-4, 5 mg
SDP
Catalog No. 502979865
Encompass_Preferred
A potent, selective antagonist of TRPV4 channels, blocking the influx of calcium induced by the TRPV4 agonist GSK634775 with IC50 values of 2, 5, and 40 nM for rat, mouse, and human isoforms, respectively; effective in vivo, as it resolves pulmonary edema resulting from myocardial infarction in mice
Catalog No. 50-297-9865 Supplier Cayman Chemical Supplier No. 177155 MG
A potent, selective antagonist of TRPV4 channels, blocking the influx of calcium induced by the TRPV4 agonist GSK634775 with IC50 values of 2, 5, and 40 nM for rat, mouse, and human isoforms, respectively; effective in vivo, as it resolves pulmonary edema resulting from myocardial infarction in mice