Learn More
Cayman Chemical GSK256066, Purity: ≥98%, CAS: 801312-28-7, 1 mg
SDP
Catalog No. 502937451
Encompass_Preferred
A potent PDE4 inhibitor (IC50s = 4.9, 3.2, 3.8, and 1.1 pM for PDE4A, PDE4B, PDE4C, and PDE4D, respectively); >380,000-fold selective for PDE4 over PDE1-3, -5, and -6 and >2,500-fold selective over PDE7; inhibits LPS-induced TNF-α production in human peripheral blood monocytes (IC50 = 0.01 nM) and human whole blood (IC50 = 126 pM); intratracheal adminstration inhibits LPS-induced pulmonary neutrophilia in rats (ED50 = 1.1 µg/kg); inhibits ovalbumin-induced pulmonary eosinophilia in rats (ED50 = 0.4 µg/kg) and LPS-induced pulmonary neutrophilia in ferrets (ED50 = 18 µg/kg)
Catalog No. 50-293-7451 Supplier Cayman Chemical Supplier No. 280121 MG
A potent PDE4 inhibitor (IC50s = 4.9, 3.2, 3.8, and 1.1 pM for PDE4A, PDE4B, PDE4C, and PDE4D, respectively); >380,000-fold selective for PDE4 over PDE1-3, -5, and -6 and >2,500-fold selective over PDE7; inhibits LPS-induced TNF-α production in human peripheral blood monocytes (IC50 = 0.01 nM) and human whole blood (IC50 = 126 pM); intratracheal adminstration inhibits LPS-induced pulmonary neutrophilia in rats (ED50 = 1.1 µg/kg); inhibits ovalbumin-induced pulmonary eosinophilia in rats (ED50 = 0.4 µg/kg) and LPS-induced pulmonary neutrophilia in ferrets (ED50 = 18 µg/kg)