Learn More
Cayman Chemical GSK3179106, Purity: ≥98%, CAS: 1627856-64-7, 25 mg
SDP
Catalog No. 502933172
Encompass_Preferred
An inhibitor of RET kinase (IC50 = 0.4 nM); selective for RET kinase over VEGFR2/KDR (IC50 = 109 nM); reduces the number of abdominal contractions induced by colorectal distension in acetic acid-sensitized rats when administered at 10 mg/kg twice per day
Catalog No. 50-293-3172 Supplier Cayman Chemical Supplier No. 2402325 MG
An inhibitor of RET kinase (IC50 = 0.4 nM); selective for RET kinase over VEGFR2/KDR (IC50 = 109 nM); reduces the number of abdominal contractions induced by colorectal distension in acetic acid-sensitized rats when administered at 10 mg/kg twice per day