Learn More
Cayman Chemical GW 405833, Purity: ≥98%, CAS: 180002-83-9, 1 mg
SDP
Catalog No. 502981894
Encompass_Preferred
A partial agonist of CB2 receptors (EC50 = 0.65 nM); demonstrates anti-inflammatory and antihyperalgesic properties in animal models of inflammation and pain
Catalog No. 50-298-1894 Supplier Cayman Chemical Supplier No. 202191 MG
A partial agonist of CB2 receptors (EC50 = 0.65 nM); demonstrates anti-inflammatory and antihyperalgesic properties in animal models of inflammation and pain