Learn More
Cayman Chemical GW 406381, Purity: ≥98%, 10 mg
SDP
Catalog No. 502950563
Encompass_Preferred
A NSAID and COX-2 inhibitor (IC50 = 3 nM); selective for COX-2 over COX-1 (IC50 = >84.2 µM); reverses chronic construction injury-induced decreases in paw withdrawal thresholds in a rat model of neuropathic pain at 5 mg/kg
Catalog No. 50-295-0563 Supplier Cayman Chemical Supplier No. 3389510 MG
A NSAID and COX-2 inhibitor (IC50 = 3 nM); selective for COX-2 over COX-1 (IC50 = >84.2 µM); reverses chronic construction injury-induced decreases in paw withdrawal thresholds in a rat model of neuropathic pain at 5 mg/kg