Learn More
Cayman Chemical H3B-6527, Purity: ≥98%, CAS: 1702259-66-2, 10 mg
SDP
Catalog No. 502936675
Encompass_Preferred
An FGFR4 inhibitor (IC50 = 50s = 320, 1,290, and 1,060 nM, respectively), as well as TAOK2, JNK2, and CSF1R (IC50s = 690, >10,000, and >10,000 nM, respectively); decreases cell growth (GI50 = 25 nM) and increases caspase-3/7 activity in Hep3B HCC cells; selectively inhibits cell growth in HCC cell lines that express high levels of FGF19; reduces tumor growth in a Hep3B HCC mouse xenograft model at 100 and 300 mg/kg
Catalog No. 50-293-6675 Supplier Cayman Chemical Supplier No. 2607210 MG
An FGFR4 inhibitor (IC50 = 50s = 320, 1,290, and 1,060 nM, respectively), as well as TAOK2, JNK2, and CSF1R (IC50s = 690, >10,000, and >10,000 nM, respectively); decreases cell growth (GI50 = 25 nM) and increases caspase-3/7 activity in Hep3B HCC cells; selectively inhibits cell growth in HCC cell lines that express high levels of FGF19; reduces tumor growth in a Hep3B HCC mouse xenograft model at 100 and 300 mg/kg