Learn More
Cayman Chemical HA-15, Purity: ≥98%, CAS: 1609402-14-3, 25 mg
SDP
Catalog No. 502911204
Encompass_Preferred
An inhibitor of GRP78; binds to GRP79 in a cell-free assay and increases the expression of the genes encoding the UPR proteins EIF2AK3, IRE1α, ATF6, DDIT3, and XBP1, as well as the apoptosis-associated genes JUN and BCL2 and the autophagy marker gene LC3; decreases the viability of A375 melanoma cells (IC50 = 2.5 µM); reduces tumor volume and induces intratumor apoptosis and autophagy in an A375 mouse xenograft model at 0.7 mg/animal per day; increases osteogenesis and reduces bone loss in a mouse model of ovariectomy-induced osteoporosis
Catalog No. 50-291-1204 Supplier Cayman Chemical Supplier No. 3577525 MG
An inhibitor of GRP78; binds to GRP79 in a cell-free assay and increases the expression of the genes encoding the UPR proteins EIF2AK3, IRE1α, ATF6, DDIT3, and XBP1, as well as the apoptosis-associated genes JUN and BCL2 and the autophagy marker gene LC3; decreases the viability of A375 melanoma cells (IC50 = 2.5 µM); reduces tumor volume and induces intratumor apoptosis and autophagy in an A375 mouse xenograft model at 0.7 mg/animal per day; increases osteogenesis and reduces bone loss in a mouse model of ovariectomy-induced osteoporosis