Learn More
Cayman Chemical HAMNO, Purity: ≥95%, CAS: 138736-73-9, 10 mg
SDP
Catalog No. 502939876
Encompass_Preferred
A small molecule inhibitor of RPA; selectively binds to and inhibits DBD-F on RPA70 up to 100 µM; inhibits etoposide-induced autophosphorylation of ATR kinase and subsequent phosphorylation of RPA32 in UMSCC38 cells at 10 and 20 µM; reduces tumor growth in UMSCC11B and UMSCC38 mouse xenograft models at 1 mg/kg when used in combination with etoposide
Catalog No. 50-293-9876 Supplier Cayman Chemical Supplier No. 2763110 MG
A small molecule inhibitor of RPA; selectively binds to and inhibits DBD-F on RPA70 up to 100 µM; inhibits etoposide-induced autophosphorylation of ATR kinase and subsequent phosphorylation of RPA32 in UMSCC38 cells at 10 and 20 µM; reduces tumor growth in UMSCC11B and UMSCC38 mouse xenograft models at 1 mg/kg when used in combination with etoposide