Learn More
Cayman Chemical HAPC-Chol, Purity: ≥95%, 10 mg
SDP
Catalog No. 502935118
Encompass_Preferred
A cationic cholesterol; has been used for siRNA delivery and gene silencing in MCF-7 cells in a luciferase assay without affecting cell viability; has been used to deliver siRNA into mice via intravenous injection, resulting in HAPC-chol accumulation in the lungs
Catalog No. 50-293-5118 Supplier Cayman Chemical Supplier No. 2658410 MG
A cationic cholesterol; has been used for siRNA delivery and gene silencing in MCF-7 cells in a luciferase assay without affecting cell viability; has been used to deliver siRNA into mice via intravenous injection, resulting in HAPC-chol accumulation in the lungs