Learn More
Cayman Chemical Hecogenin, Purity: ≥80%, CAS: 467-55-0, 1 g
SDP
Catalog No. 502985274
Encompass_Preferred
A natural steroid sapogenin with diverse biological activities; decreases fMLP-induced superoxide anion formation by rat neutrophils by 23.6% (10 μM); inhibits PMA-induced MPO release from human neutrophils (1 μg/ml); inhibits the proliferation of at least seven breast cancer cell lines (IC50s = 28.7-38.2 μM); decreases the number of glutamate-induced TUNEL-positive primary rat spinal motor neurons by 11% (1 μM); decreases the mucosal lesion area of ethanol-induced gastric ulceration in mice (15 mg/kg); decreases tumor volume in an MDA-MB-231 human breast cancer mouse xenograft model (10 mg/kg three times per week)
Catalog No. 50-298-5274 Supplier Cayman Chemical Supplier No. 213721 G
A natural steroid sapogenin with diverse biological activities; decreases fMLP-induced superoxide anion formation by rat neutrophils by 23.6% (10 μM); inhibits PMA-induced MPO release from human neutrophils (1 μg/ml); inhibits the proliferation of at least seven breast cancer cell lines (IC50s = 28.7-38.2 μM); decreases the number of glutamate-induced TUNEL-positive primary rat spinal motor neurons by 11% (1 μM); decreases the mucosal lesion area of ethanol-induced gastric ulceration in mice (15 mg/kg); decreases tumor volume in an MDA-MB-231 human breast cancer mouse xenograft model (10 mg/kg three times per week)