Learn More
Cayman Chemical Hexarelin, Purity: ≥98%, CAS: 140703-51-1, 1 mg
SDP
Catalog No. 502989428
Encompass_Preferred
A peptide agonist of GHS-R; selectively binds to GHS-R over CD36 (IC50s = 15.9 and 2,080 nM, respectively); increases plasma growth hormone levels in neonatal rats at 300 µg/kg; improves left ventricular function and reduces fibrosis in a transient LAD ligation-induced mouse model of myocardial ischemia at 0.3 mg/kg per day
Catalog No. 50-298-9428 Supplier Cayman Chemical Supplier No. 307161 MG
A peptide agonist of GHS-R; selectively binds to GHS-R over CD36 (IC50s = 15.9 and 2,080 nM, respectively); increases plasma growth hormone levels in neonatal rats at 300 µg/kg; improves left ventricular function and reduces fibrosis in a transient LAD ligation-induced mouse model of myocardial ischemia at 0.3 mg/kg per day