Learn More
Cayman Chemical Histamine, Purity: ≥95%, CAS: 51-45-6, 5 g
SDP
Catalog No. 502946227
Encompass_Preferred
A biogenic amine; activates histamine H1-4 receptors to regulate allergic and inflammatory responses, gastric acid secretion, arousal, cognition, and immune cell chemotaxis
Catalog No. 50-294-6227 Supplier Cayman Chemical Supplier No. 338285 G
A biogenic amine; activates histamine H1-4 receptors to regulate allergic and inflammatory responses, gastric acid secretion, arousal, cognition, and immune cell chemotaxis