Learn More
Cayman Chemical HNMPA-(AM)3, Purity: ≥95%, CAS: 120944-03-8, 10 mg
SDP
Catalog No. 502987153
Encompass_Preferred
A cell-permeable prodrug form of HNMPA; inhibits IRTK autophosphorylation in CHO cells expressing the human receptor in a concentration- and time-dependent manner; inhibits 2-deoxyglucose uptake but does not affect insulin-stimulated thymidine uptake in CHO cells (≤100 μM)
Catalog No. 50-298-7153 Supplier Cayman Chemical Supplier No. 2324510 MG
A cell-permeable prodrug form of HNMPA; inhibits IRTK autophosphorylation in CHO cells expressing the human receptor in a concentration- and time-dependent manner; inhibits 2-deoxyglucose uptake but does not affect insulin-stimulated thymidine uptake in CHO cells (≤100 μM)