Learn More
Cayman Chemical ANTIBODY HPPD-Q 500ug
SDP
Supplier:  Cayman Chemical 40613500 UG
Encompass_Preferred
A substituted p-phenylenediamine quinone; a derivative of 6-PPD-Q; not toxic to rainbow trout (O. mykiss) at concentrations up to 50 UG/L for 96 hours
Catalog No. 50-294-4897