Learn More
Cayman Chemical HT-2 Toxin, Purity: ≥98%, CAS: 26934-87-2, 5 mg
SDP
Catalog No. 502982548
Encompass_Preferred
A mycotoxin and an active metabolite of T-2 toxin; reduces viability of a variety of cancer cells (IC50s = 3.1-23 ng/ml) and HUVECs (IC50 = 56.4 ng/ml); induces oxidative stress, DNA damage, and autophagy in, as well as halts the development of, cultured mouse embryos at 10 nM
Catalog No. 50-298-2548 Supplier Cayman Chemical Supplier No. 204315 MG
A mycotoxin and an active metabolite of T-2 toxin; reduces viability of a variety of cancer cells (IC50s = 3.1-23 ng/ml) and HUVECs (IC50 = 56.4 ng/ml); induces oxidative stress, DNA damage, and autophagy in, as well as halts the development of, cultured mouse embryos at 10 nM