Learn More
Cayman Chemical I-BET726, Purity: ≥98%, CAS: 1300031-52-0, 1 mg
SDP
Catalog No. 502980604
Encompass_Preferred
A selective inhibitor of BET family proteins that binds BRD2, BRD3, and BRD4 with high affinity (IC50s = 41, 31, and 22 nM, respectively); inhibits cell growth and induces cytotoxicity in neuroblastoma cell lines, in vitro and in vivo
Catalog No. 50-298-0604 Supplier Cayman Chemical Supplier No. 168721 MG
A selective inhibitor of BET family proteins that binds BRD2, BRD3, and BRD4 with high affinity (IC50s = 41, 31, and 22 nM, respectively); inhibits cell growth and induces cytotoxicity in neuroblastoma cell lines, in vitro and in vivo