Learn More
Cayman Chemical IAA-94, Purity: ≥95%, CAS: 54197-31-8, 25 mg
SDP
Catalog No. 502964880
Encompass_Preferred
An IAA inhibitor of chloride channels that binds channels in bovine kidney cortex microsomes with a Ki value of 1 µM; reversibly inhibits CLIC proteins at 10 µM
Catalog No. 50-296-4880 Supplier Cayman Chemical Supplier No. 1787725 MG
An IAA inhibitor of chloride channels that binds channels in bovine kidney cortex microsomes with a Ki value of 1 µM; reversibly inhibits CLIC proteins at 10 µM