Learn More
Cayman Chemical iBET-BD2, Purity: ≥98%, CAS: 2474876-09-8, 10 mg
SDP
Catalog No. 502990729
Encompass_Preferred
A BD2 bromodomain inhibitor; inhibits BD2 in BRD2, BRD3, BRD4, and BRDT (IC50s = 264, 98, 49, and 214 nM, respectively) and is selective for BD2 over BD1 (IC50s = 10.96, 36.31, 70.55, and >50.11 µM for BRD2, BRD3, BRD4, and BDT, respectively) in a TR-FRET assay; inhibits IFN-γ-induced protein expression of MHC class I in K562 cells at 1 µM; reduces the production of anti-KLH IgM antibodies in mice immunized with KLH at 40 mg/kg
Catalog No. 50-299-0729 Supplier Cayman Chemical Supplier No. 3176610 MG
A BD2 bromodomain inhibitor; inhibits BD2 in BRD2, BRD3, BRD4, and BRDT (IC50s = 264, 98, 49, and 214 nM, respectively) and is selective for BD2 over BD1 (IC50s = 10.96, 36.31, 70.55, and >50.11 µM for BRD2, BRD3, BRD4, and BDT, respectively) in a TR-FRET assay; inhibits IFN-γ-induced protein expression of MHC class I in K562 cells at 1 µM; reduces the production of anti-KLH IgM antibodies in mice immunized with KLH at 40 mg/kg