Learn More
Cayman Chemical ICAAc, Purity: ≥98%, 25 mg
SDP
Catalog No. 502938764
Encompass_Preferred
A solvatochromic fluorescent pH probe; ex/em = 466/553, 431/515, and 418/503 nm in water, dioxane, and hexane, respectively; abs/em = 470/554 and 428/553 nm at pH 3 and 11, respectively, in aqueous buffer, with fluorescence intensity increasing as pH decreases; can be used for live cell applications
Catalog No. 50-293-8764 Supplier Cayman Chemical Supplier No. 2856425 MG
A solvatochromic fluorescent pH probe; ex/em = 466/553, 431/515, and 418/503 nm in water, dioxane, and hexane, respectively; abs/em = 470/554 and 428/553 nm at pH 3 and 11, respectively, in aqueous buffer, with fluorescence intensity increasing as pH decreases; can be used for live cell applications