Learn More
Cayman Chemical ICI 63197, Purity: ≥98%, CAS: 27277-00-5, 500 mg
SDP
Catalog No. 502910097
Encompass_Preferred
A PDE3 and PDE4 inhibitor (Kis = 9 and 10 µM, respectively); selective for PDE3 and PDE4 over PDE1 and PDE2 (Kis = 487 and >1,000 µM, respectively); inhibits acetylcholine-induced bronchospasm in isolated guinea pig lungs and histamine-induced bronchospasm in conscious guinea pigs; decreases motor activity and rectal temperature and induces forepaw shaking, head twitches, and grooming in rats
Catalog No. 50-291-0097 Supplier Cayman Chemical Supplier No. 35538500 MG
A PDE3 and PDE4 inhibitor (Kis = 9 and 10 µM, respectively); selective for PDE3 and PDE4 over PDE1 and PDE2 (Kis = 487 and >1,000 µM, respectively); inhibits acetylcholine-induced bronchospasm in isolated guinea pig lungs and histamine-induced bronchospasm in conscious guinea pigs; decreases motor activity and rectal temperature and induces forepaw shaking, head twitches, and grooming in rats