Learn More
Cayman Chemical IEM 1460, Purity: ≥98%, CAS: 121034-89-7, 10 mg
SDP
Catalog No. 502973163
Encompass_Preferred
An adamantine derivative that selectively blocks Ca2+-permeable glutamate receptor channels (IC50 = 10 μM-0.1 mM)
Catalog No. 50-297-3163 Supplier Cayman Chemical Supplier No. 1562310 MG
An adamantine derivative that selectively blocks Ca2+-permeable glutamate receptor channels (IC50 = 10 μM-0.1 mM)