Learn More
Cayman Chemical Illudin M, Purity: ≥85%, CAS: 1146-04-9, 5 mg
SDP
Catalog No. 502987096
Encompass_Preferred
A DNA alkylating agent from the fungus O. illudens; cytotoxic to HL-60 cells at 6-100 nM
Catalog No. 50-298-7096 Supplier Cayman Chemical Supplier No. 220625 MG
A DNA alkylating agent from the fungus O. illudens; cytotoxic to HL-60 cells at 6-100 nM